Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp184 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751710.3 | XP_003751758.1 | 4 | 566 | ||
XM_006254113.2 | XP_006254175.1 | 4 | 633 | |||
XM_006254114.2 | XP_006254176.1 | 4 | 758 | |||
XM_006254118.2 | XP_006254180.1 | 1 | 458 | |||
XM_008771768.1 | XP_008769990.1 | 4 | 758 | |||
GenBank mRNA | BC128784.1 | - | 395 | 7381364 |