Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCOLCE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127640.1 | NP_001121112.1 | 259 | |||
GenBank mRNA | BC161891.1 | - | 259 | 7189256 |