Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgals1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019904.1 | NP_063969.1 | 3 | 211 | ||
GenBank mRNA | BC058476.1 | - | 3 | 241 | 6920799 | |
FQ217640.1 | - | 3 | 238 | |||
FQ221017.1 | - | 3 | 237 | |||
FQ221731.1 | - | 3 | 237 | |||
FQ221808.1 | - | 3 | 238 | |||
FQ221945.1 | - | 3 | 238 | |||
FQ222079.1 | - | 3 | 238 | |||
FQ222712.1 | - | 3 | 238 | |||
FQ223159.1 | - | 3 | 237 | |||
FQ223277.1 | - | 3 | 238 | |||
FQ223897.1 | - | 3 | 238 | |||
FQ224769.1 | - | 3 | 238 | |||
FQ228416.1 | - | 2 | 165 | |||
FQ228452.1 | - | 3 | 238 | |||
FQ228459.1 | - | 3 | 237 | |||
FQ228622.1 | - | 3 | 236 | |||
FQ230068.1 | - | 3 | 238 | |||
M19036.1 | - | 3 | 211 |