Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HBA2 SIRNA Assays ›
Gene Symbol:
Hba2
Hba1
Gene Name:
hemoglobin, alpha 2
hemoglobin, alpha 1
Gene Aliases:
-
HBAM, Hba-a1, Hba-a2
Chromosome Location:
Chr.10: 15589364 - 15590207 on Build Rnor_6.0
Chromosome Location:
Chr.10: 15602794 - 15603649 on Build Rnor_6.0
UniGene ID:
Rn.203003
Rn.107334
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID
s234980
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Hba2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001007722.1
NP_001007723.1
98
GenBank mRNA
FQ209950.1
-
1
96
FQ210108.1
-
1
96
FQ210256.1
-
1
96
FQ210264.1
-
1
96
FQ210372.1
-
1
101
FQ210420.1
-
1
96
FQ210570.1
-
1
96
FQ210694.1
-
1
96
FQ211180.1
-
1
96
FQ211270.1
-
1
96
FQ211366.1
-
1
96
FQ211672.1
-
1
96
FQ211887.1
-
1
96
FQ213126.1
-
1
96
FQ214258.1
-
1
96
FQ215087.1
-
1
96
FQ217874.1
-
1
96
FQ218140.1
-
1
98
FQ218297.1
-
1
102
FQ218784.1
-
1
95
FQ221489.1
-
1
96
FQ222342.1
-
1
96
FQ222639.1
-
1
95
FQ222693.1
-
1
94
FQ223754.1
-
1
98
FQ223838.1
-
1
122
FQ224218.1
-
1
105
FQ224339.1
-
1
98
FQ224679.1
-
1
96
FQ225311.1
-
1
96
FQ225451.1
-
1
96
FQ225711.1
-
1
96
FQ225789.1
-
1
98
FQ226185.1
-
1
96
FQ226226.1
-
251
FQ226238.1
-
1
86
FQ226556.1
-
1
96
FQ226574.1
-
1
96
FQ226576.1
-
1
96
FQ226594.1
-
1
96
FQ226619.1
-
1
96
FQ226927.1
-
1
96
FQ228017.1
-
1
98
FQ228292.1
-
1
96
FQ228298.1
-
1
96
FQ228306.1
-
1
96
FQ228588.1
-
1
96
FQ229556.1
-
1
96
FQ230499.1
-
1
96
FQ230501.1
-
1
96
FQ230536.1
-
1
96
FQ230819.1
-
1
98
FQ230868.1
-
1
96
FQ230877.1
-
1
98
FQ231222.1
-
1
96
FQ231239.1
-
1
91
FQ231246.1
-
1
96
FQ231250.1
-
1
96
FQ231268.1
-
1
95
FQ231572.1
-
1
96
FQ231902.1
-
1
96
FQ232213.1
-
1
96
FQ234805.1
-
1
96
FQ235117.1
-
1
97
Gene Symbol: Hba1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_013096.1
NP_037228.1
1
98
GenBank mRNA
BC059150.1
-
1
97
6918464
BC091567.1
-
85
7458326
BC160818.1
-
1
81
7375173
FQ209733.1
-
1
98
FQ209744.1
-
1
98
FQ209909.1
-
1
98
FQ209912.1
-
1
98
FQ210001.1
-
1
98
FQ210150.1
-
1
96
FQ210186.1
-
1
96
FQ210277.1
-
1
96
FQ210349.1
-
1
96
FQ210635.1
-
1
98
FQ210643.1
-
1
96
FQ210690.1
-
1
97
FQ210723.1
-
1
96
FQ210862.1
-
1
96
FQ210866.1
-
1
96
FQ211077.1
-
1
98
FQ211217.1
-
1
96
FQ211220.1
-
1
98
FQ211232.1
-
1
97
FQ211239.1
-
1
96
FQ211351.1
-
1
96
FQ211374.1
-
1
96
FQ211431.1
-
1
96
FQ211513.1
-
1
96
FQ211520.1
-
1
96
FQ211755.1
-
1
96
FQ211911.1
-
1
96
FQ211972.1
-
1
96
FQ212025.1
-
1
98
FQ212144.1
-
1
98
FQ212153.1
-
1
98
FQ212186.1
-
1
96
FQ212232.1
-
1
98
FQ212457.1
-
1
96
FQ212854.1
-
1
96
FQ213301.1
-
1
96
FQ214066.1
-
1
96
FQ214223.1
-
1
96
FQ216243.1
-
1
96
FQ217723.1
-
1
96
FQ217724.1
-
1
96
FQ217869.1
-
1
96
FQ217988.1
-
1
98
FQ218117.1
-
1
96
FQ218146.1
-
96
FQ218612.1
-
1
95
FQ218968.1
-
1
98
FQ219715.1
-
1
96
FQ220786.1
-
1
96
FQ221656.1
-
1
97
FQ221912.1
-
1
98
FQ221952.1
-
1
96
FQ222431.1
-
1
96
FQ222454.1
-
1
96
FQ222672.1
-
1
96
FQ222750.1
-
1
98
FQ223017.1
-
1
97
FQ224581.1
-
1
96
FQ225427.1
-
1
97
FQ225429.1
-
1
98
FQ225440.1
-
1
97
FQ225452.1
-
1
96
FQ225777.1
-
1
96
FQ225808.1
-
1
99
FQ226118.1
-
1
98
FQ226173.1
-
1
98
FQ226175.1
-
1
96
FQ226203.1
-
1
96
FQ226615.1
-
1
96
FQ227082.1
-
1
96
FQ228301.1
-
1
96
FQ228436.1
-
96
FQ230483.1
-
1
98
FQ230901.1
-
1
96
FQ231201.1
-
1
88
FQ231290.1
-
1
96
FQ232272.1
-
1
96
FQ232481.1
-
1
96
FQ234683.1
-
1
96
M17083.1
-
1
98
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
positive regulation of cell death
response to hydrogen peroxide
hydrogen peroxide catabolic process
erythrocyte development
protein heterooligomerization
oxidation-reduction process
oxygen transport
negative regulation of blood pressure
regulation of sensory perception of pain
peroxidase activity
haptoglobin binding
beta-amyloid binding
oxygen transporter activity
iron ion binding
protein binding
oxygen binding
heme binding
organic acid binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0