Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom2r65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099654.1 | NP_001093124.1 | 386 |
Gene Symbol: LOC100911926 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766128.1 | XP_008764350.1 | 2 | 430 |
Set Membership: |
G-protein coupled receptor |