Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bloc1s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105941.2 | NP_001099411.2 | 3 | 342 | ||
GenBank mRNA | CB613801.1 | - | 3 | 348 | ||
FQ217708.1 | - | 3 | 355 | |||
FQ221841.1 | - | 3 | 302 |