Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il36b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108570.1 | NP_001102040.1 | 3 | 349 | ||
XM_006233614.1 | XP_006233676.1 | 4 | 488 | |||
XM_006233615.1 | XP_006233677.1 | 5 | 1808 | |||
XM_006233616.2 | XP_006233678.1 | 5 | 691 | |||
XM_006233617.2 | XP_006233679.1 | 5 | 904 |