Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WSCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015253.1 | NP_056068.1 | 9 | 1717 | ||
XM_005256572.1 | XP_005256629.1 | 9 | 1805 | |||
XM_005256573.1 | XP_005256630.1 | 9 | 1780 | |||
GenBank mRNA | AB011095.1 | - | 8 | 1069 | ||
AK000243.1 | - | 7 | 1190 | |||
AK289603.1 | - | 9 | 1776 | |||
AK309645.1 | - | 8 | 1169 | |||
AL833730.1 | - | 9 | 1717 | |||
BC009975.2 | - | 9 | 1742 | 4110708 |