Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109331.2 | NP_001102801.1 | 2 | 242 |
Gene Symbol: LOC100910177 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751511.3 | XP_003751559.2 | 1 | 231 |