Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184801.1 | NP_001171730.1 | 6, 7 | 1109 | ||
NM_015255.2 | NP_056070.1 | 6, 7 | 1109 | |||
XM_005248965.4 | XP_005249022.1 | 6, 7 | 1226 | |||
XM_005248966.3 | XP_005249023.1 | 5, 6 | 513 | |||
XM_011514438.2 | XP_011512740.2 | 6, 7 | 1309 | |||
XM_017010594.1 | XP_016866083.1 | 6, 7 | 1310 | |||
XM_017010595.1 | XP_016866084.1 | 6, 7 | 1310 | |||
XM_017010596.1 | XP_016866085.1 | 6, 7 | 1144 | |||
XM_017010597.1 | XP_016866086.1 | 6, 7 | 1311 | |||
GenBank mRNA | AY061884.1 | - | 6, 7 | 811 | ||
BC024217.2 | - | 6, 7 | 1088 | |||
BC064512.1 | - | 6, 7 | 1088 | |||
BX647467.1 | - | 1, 2 | 91 |