Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lsm6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126085.1 | NP_001119557.1 | 4 | 351 | ||
XM_006255408.1 | XP_006255470.1 | 3 | 266 |
Gene Symbol: LOC100360750 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002728002.4 | XP_002728048.1 | 1 | 324 |