Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACSL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009185.2 | NP_001009185.1 | 3 | 432 | ||
NM_001205247.1 | NP_001192176.1 | 3 | 327 | |||
NM_001205248.1 | NP_001192177.1 | 3 | 419 | |||
NM_001205250.1 | NP_001192179.1 | 3 | 429 | |||
NM_001205251.1 | NP_001192180.1 | 3 | 314 | |||
NM_015256.3 | NP_056071.2 | 3 | 432 | |||
GenBank mRNA | AB020644.1 | - | 3 | 392 | ||
AF102845.1 | - | 2 | 209 | |||
AF129166.1 | - | 3 | 362 | |||
AK294291.1 | - | 3 | 302 | |||
AK314452.1 | - | 3 | 334 | |||
BC026161.1 | - | 3 | 328 | 4830357 | ||
BC047453.1 | - | 3 | 236 | 5270879 | ||
BI464352.1 | - | 3 | 415 | |||
BI545119.1 | - | 3 | 433 | |||
BI548729.1 | - | 3 | 432 | |||
DA200154.1 | - | 3 | 442 | |||
DA801862.1 | - | 3 | 429 | |||
DB461818.1 | - | 3 | 369 | |||
DQ083031.1 | - | 2 | 248 | |||
DQ655980.2 | - | 3 | 364 |