Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107075.1 | NP_001100545.1 | 5 | 644 | ||
XM_006247890.2 | XP_006247952.1 | 5 | 1041 | |||
XM_006247891.2 | XP_006247953.1 | 5 | 1041 | |||
GenBank mRNA | BC168919.1 | - | 4 | 509 |