Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100134871 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113223.1 | NP_001106694.1 | 3 | 560 | ||
GenBank mRNA | FQ209951.1 | - | 3 | 563 | ||
FQ210023.1 | - | 3 | 563 | |||
FQ210060.1 | - | 3 | 561 | |||
FQ210106.1 | - | 3 | 561 | |||
FQ210483.1 | - | 3 | 561 | |||
FQ210600.1 | - | 3 | 563 | |||
FQ210669.1 | - | 3 | 557 | |||
FQ210740.1 | - | 3 | 561 | |||
FQ211039.1 | - | 3 | 555 | |||
FQ211266.1 | - | 3 | 561 | |||
FQ211272.1 | - | 3 | 561 | |||
FQ211275.1 | - | 3 | 561 | |||
FQ211294.1 | - | 3 | 563 | |||
FQ211362.1 | - | 3 | 561 | |||
FQ211415.1 | - | 3 | 561 | |||
FQ211424.1 | - | 3 | 561 | |||
FQ211697.1 | - | 3 | 563 | |||
FQ212253.1 | - | 3 | 561 | |||
FQ212317.1 | - | 3 | 563 | |||
FQ212406.1 | - | 3 | 563 | |||
FQ212421.1 | - | 3 | 561 | |||
FQ213080.1 | - | 3 | 590 | |||
FQ214057.1 | - | 3 | 561 | |||
FQ214129.1 | - | 3 | 561 | |||
FQ217131.1 | - | 3 | 559 | |||
FQ217673.1 | - | 3 | 561 | |||
FQ218002.1 | - | 3 | 562 | |||
FQ218246.1 | - | 3 | 533 | |||
FQ218266.1 | - | 3 | 599 | |||
FQ218689.1 | - | 3 | 561 | |||
FQ218821.1 | - | 3 | 561 | |||
FQ219109.1 | - | 3 | 561 | |||
FQ219133.1 | - | 3 | 592 | |||
FQ221588.1 | - | 3 | 559 | |||
FQ222844.1 | - | 3 | 561 | |||
FQ224149.1 | - | 3 | 561 | |||
FQ224301.1 | - | 3 | 561 | |||
FQ224598.1 | - | 3 | 561 | |||
FQ224632.1 | - | 3 | 561 | |||
FQ224738.1 | - | 3 | 561 | |||
FQ224763.1 | - | 3 | 600 | |||
FQ225377.1 | - | 3 | 561 | |||
FQ225739.1 | - | 3 | 563 | |||
FQ226535.1 | - | 3 | 561 | |||
FQ226609.1 | - | 3 | 563 | |||
FQ228555.1 | - | 3 | 561 | |||
FQ230881.1 | - | 3 | 563 | |||
FQ232298.1 | - | 705 | ||||
FQ232442.1 | - | 3 | 592 | |||
FQ233001.1 | - | 3 | 561 | |||
FQ234495.1 | - | 3 | 562 | |||
X05080.1 | - | 3 | 560 | |||
X16418.1 | - | 3 | 526 |
Gene Symbol: Hbb-b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198776.1 | NP_942071.1 | 3 | 565 | ||
GenBank mRNA | BC058502.1 | - | 3 | 565 | 6887460 | |
FQ218177.1 | - | 3 | 559 | |||
M32509.1 | - | 223 |
Gene Symbol: LOC103694855 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773799.1 | XP_008772021.1 | 3 | 601 |