Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csad | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134454.1 | NP_001127926.1 | 10 | 971 | ||
NM_021750.2 | NP_068518.1 | 12 | 1048 | |||
XM_006242438.2 | XP_006242500.1 | 12 | 1457 | |||
XM_006242439.1 | XP_006242501.1 | 12 | 1054 | |||
XM_006242440.2 | XP_006242502.1 | 13 | 1533 | |||
XM_006242441.2 | XP_006242503.1 | 12 | 1115 | |||
XM_006242442.2 | XP_006242504.1 | 13 | 1148 | |||
XM_008765782.1 | XP_008764004.1 | 12 | 1235 | |||
GenBank mRNA | AJ132661.1 | - | 13 | 1153 | ||
BC081804.1 | - | 12 | 1048 | 7106840 | ||
EU786150.1 | - | 11 | 1020 | |||
EU906909.1 | - | 10 | 971 | |||
M64755.1 | - | 12 | 1087 | |||
X94152.1 | - | 12 | 1206 |