Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnl3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081958.1 | NP_001075427.1 | 14, 15 | 1492 | ||
GenBank mRNA | CB801341.1 | - | 2, 3 | 302 | ||
FQ223839.1 | - | 1130 |