Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008817.1 | NP_001008817.1 | 3 | 485 | ||
XM_006247392.2 | XP_006247454.1 | 3 | 530 |