Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIN3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297595.1 | NP_001284524.1 | 10 | 1367 | ||
NM_001297597.1 | NP_001284526.1 | 1 | 473 | |||
NM_015260.3 | NP_056075.1 | 11 | 1463 | |||
XM_006722704.1 | XP_006722767.1 | 1 | 78 | |||
GenBank mRNA | AB014600.2 | - | 10 | 1354 | ||
AK001633.1 | - | 4 | 468 | |||
AK295618.1 | - | 1 | 412 | |||
AY706204.1 | - | 11 | 1443 | |||
DC332740.1 | - | 1 | 474 |