Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130674.2 | NP_001124146.1 | 4 | 590 | ||
NM_001199671.1 | NP_001186600.1 | 5 | 1405 | |||
NM_001199672.1 | NP_001186601.1 | 5 | 1405 | |||
NM_001199673.1 | NP_001186602.1 | 4 | 590 | |||
NM_001219.4 | NP_001210.1 | 4 | 590 | |||
NR_074086.1 | 2 | 164 | ||||
XM_017012659.1 | XP_016868148.1 | 4 | 955 | |||
GenBank mRNA | AF013759.1 | - | 4 | 496 | ||
AF257659.1 | - | 3 | 381 | |||
AF345637.1 | - | 4 | 483 | |||
AK027460.1 | - | 4 | 519 | |||
AK056338.1 | - | 5 | 1293 | |||
AK074862.1 | - | 4 | 540 | |||
AK312446.1 | - | 4 | 533 | |||
BC013383.2 | - | 4 | 489 | 3507961 | ||
BT006826.1 | - | 3 | 438 | |||
BT020143.1 | - | 3 | 438 | |||
CR457039.1 | - | 3 | 438 | |||
CR541835.1 | - | 3 | 438 | |||
HM002604.1 | - | 5 | 1406 | |||
HM002605.1 | - | 5 | 1406 | |||
HM002606.1 | - | 4 | 590 | |||
HM002607.1 | - | 4 | 590 | |||
HM002609.1 | - | 4 | 590 | |||
HM002611.1 | - | 4 | 590 | |||
HM002613.1 | - | 3 | 395 | |||
HM002615.1 | - | 6 | 1600 | |||
HM002616.1 | - | 2 | 164 | |||
U67280.1 | - | 4 | 500 |