Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defb12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037514.1 | NP_001032603.1 | 1 | 56 | ||
GenBank mRNA | AY621344.1 | - | 1 | 56 |
Gene Symbol: LOC100912990 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771371.1 | XP_008769593.1 | 601 |