Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ces1f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103359.2 | NP_001096829.2 | 14 | 1643 | ||
GenBank mRNA | BC128711.1 | - | 14 | 1625 | 7368807 | |
U10697.1 | - | 14 | 1623 | |||
X81825.1 | - | 14 | 1622 |