Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myl6l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100983.1 | NP_001094453.1 | 3 | 341 | ||
GenBank mRNA | S77858.1 | - | 3 | 341 |
Gene Symbol: Myl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109484.1 | NP_001102954.1 | 4 | 397 | ||
XM_008765039.1 | XP_008763261.1 | 4 | 391 | |||
XM_008765040.1 | XP_008763262.1 | 4 | 391 | |||
GenBank mRNA | BC166586.1 | - | 4 | 362 | 7934414 | |
FQ217612.1 | - | 4 | 358 | |||
FQ217952.1 | - | 4 | 370 | |||
FQ221005.1 | - | 4 | 368 | |||
FQ221192.1 | - | 4 | 364 | |||
FQ221499.1 | - | 4 | 366 | |||
FQ221709.1 | - | 4 | 398 | |||
FQ221718.1 | - | 4 | 369 | |||
FQ221751.1 | - | 4 | 365 | |||
FQ221963.1 | - | 4 | 366 | |||
FQ222160.1 | - | 4 | 367 | |||
FQ222384.1 | - | 4 | 369 | |||
FQ222504.1 | - | 4 | 346 | |||
FQ223317.1 | - | 4 | 372 | |||
FQ228533.1 | - | 4 | 369 |