Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Havcr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100762.1 | NP_001094232.1 | 6, 7 | 759 | ||
XM_006246171.2 | XP_006246233.1 | 5, 6 | 671 |