Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199671.1 | NP_001186600.1 | 4 | 1286 | ||
NM_001199673.1 | NP_001186602.1 | 3 | 471 | |||
NM_001219.4 | NP_001210.1 | 3 | 471 | |||
XM_017012659.1 | XP_016868148.1 | 3 | 836 | |||
GenBank mRNA | AF013759.1 | - | 3 | 377 | ||
AK056338.1 | - | 4 | 1174 | |||
AK074862.1 | - | 3 | 421 | |||
AK312446.1 | - | 3 | 414 | |||
BC013383.2 | - | 3 | 370 | 3507961 | ||
BT006826.1 | - | 2 | 319 | |||
BT020143.1 | - | 2 | 319 | |||
CR457039.1 | - | 2 | 319 | |||
CR541835.1 | - | 2 | 319 | |||
DA023987.1 | - | 3 | 471 | |||
HM002604.1 | - | 4 | 1287 | |||
HM002606.1 | - | 3 | 471 | |||
HM002607.1 | - | 3 | 471 | |||
HM002611.1 | - | 3 | 471 | |||
HM002615.1 | - | 4 | 1287 | |||
U67280.1 | - | 3 | 381 |