Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl35a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021264.3 | NP_067087.1 | 275 | |||
GenBank mRNA | X03475.1 | - | 275 |
Gene Symbol: LOC100359498 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002727895.4 | XP_002727941.1 | 4 | 366 | ||
XM_002727896.4 | XP_002727942.1 | 4 | 371 | |||
GenBank mRNA | FQ210925.1 | - | 4 | 335 |