Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC9A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001260491.1 | NP_001247420.1 | 7 | 800 | ||
NM_015266.2 | NP_056081.1 | 6, 7 | 752 | |||
XM_006723756.1 | XP_006723819.1 | 6, 7 | 614 | |||
XM_011528736.1 | XP_011527038.1 | 7 | 662 | |||
XM_011528737.1 | XP_011527039.1 | 6 | 535 | |||
XM_011528738.1 | XP_011527040.1 | 7 | 662 | |||
XM_011528739.1 | XP_011527041.1 | 7 | 662 | |||
XM_011528740.1 | XP_011527042.1 | 7 | 662 | |||
XM_011528741.1 | XP_011527043.1 | 7 | 662 | |||
XM_011528745.1 | XP_011527047.1 | 7 | 662 | |||
XM_017027754.1 | XP_016883243.1 | 7 | 662 | |||
XM_017027756.1 | XP_016883245.1 | 7 | 606 | |||
GenBank mRNA | AB023156.2 | - | 6, 7 | 584 | ||
AK300318.1 | - | 7 | 685 | |||
AK310530.1 | - | 6, 7 | 554 | |||
BC112213.1 | - | 6, 7 | 574 | 8327681 | ||
BX430866.2 | - | 1, 2 | 78 | |||
CR749638.1 | - | 6, 7 | 629 |