Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLC9A8 SIRNA Assays ›
Gene Symbol:
SLC9A8
Gene Name:
solute carrier family 9 member A8
Gene Aliases:
NHE-8, NHE8
Chromosome Location:
Chr.20: 49812713 - 49892242 on Build GRCh38
UniGene ID:
Hs.444202
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s23544
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SLC9A8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001260491.1
NP_001247420.1
12, 13
1420
NM_015266.2
NP_056081.1
12, 13
1372
NR_048537.1
11, 12
1337
NR_048538.1
10, 11
1253
NR_048539.1
10, 11
1209
XM_006723756.1
XP_006723819.1
13, 14
1333
XM_011528736.1
XP_011527038.1
13, 14
1381
XM_011528737.1
XP_011527039.1
12, 13
1254
XM_011528738.1
XP_011527040.1
12, 13
1264
XM_011528739.1
XP_011527041.1
12, 13
1258
XM_011528740.1
XP_011527042.1
13, 14
1381
XM_011528741.1
XP_011527043.1
11, 12
1165
XM_011528742.1
XP_011527044.1
11, 12
1154
XM_011528743.2
XP_011527045.1
5, 6
518
XM_011528744.2
XP_011527046.1
4, 5
349
XM_017027754.1
XP_016883243.1
13, 14
1381
XM_017027755.1
XP_016883244.1
12, 13
1242
XM_017027756.1
XP_016883245.1
13, 14
1330
GenBank mRNA
AB023156.2
-
12, 13
1204
AK295665.1
-
10, 11
1055
AK295803.1
-
11, 12
1012
AK295822.1
-
11, 12
1130
AK300318.1
-
12, 13
1305
AK310530.1
-
12, 13
1174
AL833574.1
-
1, 2
71
BC112213.1
-
12, 13
1194
8327681
BX430866.2
-
7, 8
698
CR749638.1
-
12, 13
1249
Back To Top
More Information
Panther Classification:
Molecular Function -
transporter
secondary carrier transporter
Gene Ontology Categories:
Function(s)
Process(es)
ion transport
regulation of intracellular pH
potassium ion transmembrane transport
sodium ion import across plasma membrane
hydrogen ion transmembrane transport
sodium:proton antiporter activity
potassium:proton antiporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-slhn9:80/100.66.78.111:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline