Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk2ap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109498.1 | NP_001102968.1 | 2 | 220 | ||
XM_006230902.2 | XP_006230964.1 | 2 | 610 | |||
XM_006230903.2 | XP_006230965.1 | 2 | 158 | |||
GenBank mRNA | FQ234216.1 | - | 2 | 222 |