Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trmt112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106330.2 | NP_001099800.1 | 4 | 463 | ||
GenBank mRNA | BC166890.1 | - | 4 | 447 | 6918473 | |
BC168731.1 | - | 4 | 447 | 6918571 | ||
BC168762.1 | - | 4 | 447 | 6889581 | ||
BC168764.1 | - | 4 | 447 | 6889585 | ||
BP485075.1 | - | 4 | 466 | |||
DV720088.1 | - | 4 | 656 | |||
FQ213514.1 | - | 4 | 795 |