Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif3k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106242.1 | NP_001099712.1 | 2 | 201 | ||
XM_006228715.2 | XP_006228777.1 | 5 | 508 |