Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015267.3 | NP_056082.2 | 9, 10 | 900 | ||
XM_011538061.2 | XP_011536363.1 | 9, 10 | 2410 | |||
XM_011538063.2 | XP_011536365.1 | 8, 9 | 2275 | |||
XM_011538069.2 | XP_011536371.1 | 9, 10 | 749 | |||
XM_011538070.2 | XP_011536372.1 | 9, 10 | 1659 | |||
XM_017019080.1 | XP_016874569.1 | 10, 11 | 1742 | |||
XM_017019081.1 | XP_016874570.1 | 10, 11 | 805 | |||
GenBank mRNA | AB006631.2 | - | 9, 10 | 899 | ||
BC151245.1 | - | 9, 10 | 910 | 8860519 |