Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kitlg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021843.4 | NP_068615.1 | 7, 8 | 882 | ||
NM_021844.2 | NP_068616.1 | 6, 7 | 798 | |||
XM_008765339.1 | XP_008763561.1 | 7, 8 | 926 | |||
XM_008765340.1 | XP_008763562.1 | 6, 7 | 842 | |||
GenBank mRNA | AB105879.1 | - | 6, 7 | 798 | ||
AF071204.1 | - | 7, 8 | 717 | |||
AF071205.1 | - | 6, 7 | 634 |