Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1453 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000772.1 | NP_001000772.1 | 1 | 258 |
Gene Symbol: Olr1451 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000957.1 | NP_001000957.1 | 258 |
Gene Symbol: Olr1455 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000771.1 | NP_001000771.1 | 1 | 258 |