Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015268.3 | NP_056083.3 | 39 | 4576 | ||
XM_005247245.4 | XP_005247302.1 | 40 | 4773 | |||
XM_017006036.1 | XP_016861525.1 | 26 | 3033 | |||
GenBank mRNA | AB014578.2 | - | 39 | 4625 | ||
AK023168.1 | - | 8 | 1004 | |||
AK125330.1 | - | 16 | 1960 | |||
AK127112.1 | - | 23 | 2664 | |||
AY779857.1 | - | 39 | 4576 | |||
BC151246.1 | - | 40 | 4646 | 9007477 |