Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam219a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109616.1 | NP_001103086.1 | 5, 6 | 627 | ||
XM_006238100.2 | XP_006238162.1 | 5, 6 | 837 | |||
XM_006238102.2 | XP_006238164.1 | 5, 6 | 784 |