Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hist1h2bd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108414.1 | NP_001101884.1 | 1 | 118 |
Gene Symbol: Hist1h2bcl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771769.1 | XP_008769991.1 | 1 | 182 |
Gene Symbol: Hist1h2bf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_225384.5 | XP_225384.1 | 1 | 128 |
Gene Symbol: Hist1h2bk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109400.1 | NP_001102870.1 | 1 | 118 |
Gene Symbol: Hist1h2bl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022647.1 | NP_072173.1 | 1 | 118 |
Gene Symbol: Hist1h2bo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106114.1 | NP_001099584.1 | 1 | 118 |
Gene Symbol: LOC100359692 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006254120.2 | XP_006254182.1 | 1 | 147 |
Gene Symbol: LOC100365043 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771682.1 | XP_008769904.1 | 1 | 171 |
Gene Symbol: LOC100910200 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771772.1 | XP_008769994.1 | 1 | 176 |