Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defb11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037505.1 | NP_001032594.1 | 2 | 131 | ||
GenBank mRNA | AY621343.1 | - | 2 | 131 |