Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Minpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019263.1 | NP_062136.1 | 5 | 1199 | ||
XM_008760326.1 | XP_008758548.1 | 5 | 776 | |||
XM_008760327.1 | XP_008758549.1 | 3 | 1033 | |||
GenBank mRNA | AF012714.2 | - | 5 | 1199 | ||
CO388212.1 | - | 2 | 176 |