Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtdr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127557.2 | NP_001121029.1 | 3 | 392 | ||
XM_006256340.2 | XP_006256402.1 | 4 | 448 | |||
XM_006256341.2 | XP_006256403.1 | 4 | 501 | |||
GenBank mRNA | BC162057.1 | - | 3 | 394 | 7114691 |