Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037549.1 | NP_001032638.1 | 2 | 91 | ||
GenBank mRNA | AY621340.1 | - | 2 | 91 |