Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl23a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108283.1 | NP_001101753.1 | 3 | 296 | ||
GenBank mRNA | BC168782.1 | - | 3 | 271 | 6921912 | |
FQ210401.1 | - | 3 | 271 | |||
FQ210899.1 | - | 3 | 272 | |||
FQ210939.1 | - | 3 | 272 | |||
FQ212155.1 | - | 3 | 271 | |||
FQ213974.1 | - | 3 | 271 | |||
FQ220609.1 | - | 3 | 271 | |||
FQ221032.1 | - | 3 | 271 | |||
FQ221170.1 | - | 3 | 262 | |||
FQ222102.1 | - | 3 | 271 | |||
FQ222310.1 | - | 3 | 271 | |||
FQ222471.1 | - | 3 | 271 | |||
FQ222491.1 | - | 3 | 271 | |||
FQ222732.1 | - | 3 | 272 | |||
FQ223377.1 | - | 3 | 271 | |||
FQ224628.1 | - | 3 | 271 | |||
FQ228299.1 | - | 3 | 275 | |||
FQ228575.1 | - | 3 | 271 | |||
FQ229110.1 | - | 3 | 271 | |||
X65228.1 | - | 3 | 274 |