Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145418.1 | NP_001138890.1 | 19, 20 | 5703 | ||
XM_006724171.3 | XP_006724234.1 | 18, 19 | 14481 | |||
XM_011530018.2 | XP_011528320.1 | 19, 20 | 5882 | |||
XM_017028673.1 | XP_016884162.1 | 19, 20 | 5676 | |||
GenBank mRNA | AB028966.3 | - | 14, 15 | 4496 | ||
AB665284.1 | - | 19, 20 | 5651 | |||
BC016465.1 | - | 11, 12 | 2247 | 4154050 |