Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106270.2 | NP_001099740.2 | 3 | 470 | ||
NM_001198638.1 | NP_001185567.1 | 3 | 246 | |||
GenBank mRNA | CB767890.1 | - | 3 | 246 |