Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPY19L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015283.1 | NP_056098.1 | 14 | 1265 | ||
XM_011515245.2 | XP_011513547.1 | 14 | 1414 | |||
XM_011515246.2 | XP_011513548.1 | 13 | 1327 | |||
GenBank mRNA | AB020684.1 | - | 11 | 836 | ||
DQ287932.1 | - | 14 | 1265 |