Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Obp2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128139.2 | NP_001121611.1 | 2 | 179 | ||
XM_008761608.1 | XP_008759830.1 | 2 | 1502 | |||
GenBank mRNA | DQ537493.1 | - | 2 | 181 |