Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127638.1 | NP_001121110.1 | 10 | 1556 | ||
XM_008769764.1 | XP_008767986.1 | 10 | 1699 | |||
GenBank mRNA | BC166443.1 | - | 10 | 1556 | 8363035 |