Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem255b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106094.2 | NP_001099564.1 | 3 | 455 | ||
XM_008771387.1 | XP_008769609.1 | 9 | 1203 | |||
XM_008771388.1 | XP_008769610.1 | 8 | 1142 | |||
XM_008771389.1 | XP_008769611.1 | 8 | 1114 | |||
GenBank mRNA | BC169123.1 | - | 3 | 455 | 6887046 |