Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014786.1 | NP_001014786.1 | 1 | 227 |
Gene Symbol: Ifna11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_233150.6 | XP_233150.2 | 1 | 559 |
Gene Symbol: Ifna16l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_575856.2 | XP_575856.1 | 1 | 230 |
Gene Symbol: Ifna2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271218.1 | NP_001258147.1 | 1 | 227 |
Gene Symbol: LOC100912314 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749974.3 | XP_003750022.1 | 1 | 582 |