Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmyc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013163.2 | NP_001013181.2 | 1 | 289 | ||
GenBank mRNA | DQ191782.1 | - | 1 | 285 | ||
DQ191783.1 | - | 1 | 272 | |||
DQ191784.1 | - | 1 | 289 | |||
DQ191785.1 | - | 1 | 269 | |||
DQ191786.1 | - | 1 | 289 |