Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oaz2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109899.2 | NP_001103369.1 | 2, 3 | 353 | ||
NM_001301297.1 | NP_001288226.1 | 2, 3 | 350 | |||
GenBank mRNA | CB586487.1 | - | 2, 3 | 352 | ||
FQ211793.1 | - | 2, 3 | 353 | |||
FQ214772.1 | - | 2, 3 | 353 |